Assay Details
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002748265.4 | XP_002748311.1 | 6 - 7 | 1858 | 64 | |
XM_008997218.2 | XP_008995466.1 | 6 - 7 | 1858 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
