Assay Details
Gene Symbol: SRSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754051.4 | XP_002754097.2 | 3 - 4 | 375 | 69 | |
XM_009006244.2 | XP_009004492.1 | 3 - 4 | 372 | 69 | ||
XM_009006245.2 | XP_009004493.1 | 3 - 4 | 373 | 69 | ||
XM_009006246.2 | XP_009004494.1 | 3 - 4 | 376 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
