Assay Details
| Gene Symbol: SUCLG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757564.4 | XP_002757610.3 | 3 - 4 | 516 | 93 | |
| XM_008980422.2 | XP_008978670.1 | 3 - 4 | 495 | 93 | ||
More Information
Set Membership: |
|

SUCLG1
Genomic Map