Assay Details
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750838.3 | XP_002750884.1 | 3 - 4 | 6502 | 56 | |
XM_017974758.1 | XP_017830247.1 | 2 - 3 | 170 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017974758.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
