Assay Details
Gene Symbol: APEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009005658.2 | XP_009003906.1 | 3 - 4 | 579 | 97 | |
XM_009005659.2 | XP_009003907.1 | 3 - 4 | 387 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
