Assay Details
| Gene Symbol: PAPSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992879.1 | XP_008991127.1 | 4 - 5 | 802 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAPSS1| Gene Symbol: PAPSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992879.1 | XP_008991127.1 | 4 - 5 | 802 | 98 | |
Set Membership: |
|