Assay Details
Gene Symbol: MBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757252.4 | XP_002757298.2 | 3 - 4 | 578 | 67 | |
XM_017963478.1 | XP_017818967.1 | 3 - 4 | 578 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
