Assay Details
| Gene Symbol: C13H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757205.4 | XP_002757251.1 | 2 - 3 | 1081 | 67 | |
| XM_017963450.1 | XP_017818939.1 | 2 - 3 | 918 | 67 | ||
More Information
Set Membership: |
|

C13H18orf25
Genomic Map