Assay Details
Gene Symbol: ERICH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017965758.1 | XP_017821247.1 | 12 - 13 | 2210 | 67 | |
XM_017965759.1 | XP_017821248.1 | 13 - 14 | 2057 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
