Assay Details
| Gene Symbol: PARP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981755.2 | XP_008980003.1 | 6 - 7 | 1235 | 53 | |
| XM_008981756.2 | XP_008980004.1 | 6 - 7 | 1261 | 53 | ||
| XM_008981757.2 | XP_008980005.1 | 6 - 7 | 1175 | 53 | ||
| XM_008981758.1 | XP_008980006.1 | 6 - 7 | 1428 | 53 | ||
| XM_008981759.2 | XP_008980007.1 | 6 - 7 | 1170 | 53 | ||
| XM_008981760.2 | XP_008980008.1 | 6 - 7 | 1428 | 53 | ||
More Information
Set Membership: |
|

PARP3
Genomic Map