Assay Details
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758184.3 | XP_002758230.2 | 5 - 6 | 1162 | 85 | |
| XM_008981384.2 | XP_008979632.1 | 8 - 9 | 1289 | 85 | ||
| XM_008981385.2 | XP_008979633.1 | 7 - 8 | 1222 | 85 | ||
| XM_008981386.2 | XP_008979634.1 | 7 - 8 | 1214 | 85 | ||
| XM_008981388.2 | XP_008979636.1 | 7 - 8 | 1221 | 85 | ||
| XM_008981389.2 | XP_008979637.1 | 5 - 6 | 1151 | 85 | ||
| XM_017964771.1 | XP_017820260.1 | 5 - 6 | 1165 | 85 | ||
| XM_017964772.1 | XP_017820261.1 | 7 - 8 | 1222 | 85 | ||
| XM_017964773.1 | XP_017820262.1 | 5 - 6 | 1159 | 85 | ||
More Information
Set Membership: |
|

TP63
Genomic Map