Assay Details
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017966555.1 | XP_017822044.1 | 3 - 4 | 1629 | 62 | |
XM_017966556.1 | XP_017822045.1 | 3 - 4 | 1627 | 62 | ||
XM_017966557.1 | XP_017822046.1 | 3 - 4 | 1625 | 62 | ||
XM_017966558.1 | XP_017822047.1 | 4 - 5 | 1303 | 62 | ||
XM_017966559.1 | XP_017822048.1 | 4 - 5 | 1300 | 62 | ||
XM_017966560.1 | XP_017822049.1 | 4 - 5 | 1303 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
