Assay Details
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008985481.2 | XP_008983729.1 | 8 - 9 | 1087 | 62 | |
GenBank mRNA | AJ132342.1 | - | 8 - 9 | 1040 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
