Assay Details
| Gene Symbol: CDS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732474.2 | XP_003732522.2 | 12 - 13 | 1786 | 118 | |
| XM_017970451.1 | XP_017825940.1 | 11 - 12 | 1666 | 118 | ||
More Information
Set Membership: |
|

CDS1
Genomic Map