Assay Details
| Gene Symbol: PIP5K1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759939.4 | XP_002759985.3 | 4 - 5 | 740 | 73 | |
| XM_008984335.2 | XP_008982583.1 | 4 - 5 | 740 | 73 | ||
| XM_008984337.2 | XP_008982585.1 | 4 - 5 | 737 | 73 | ||
| XM_008984338.2 | XP_008982586.1 | 3 - 4 | 701 | 73 | ||
| XM_008984339.2 | XP_008982587.1 | 3 - 4 | 698 | 73 | ||
| XM_017966258.1 | XP_017821747.1 | 4 - 5 | 740 | 73 | ||
| XM_017966259.1 | XP_017821748.1 | 3 - 4 | 698 | 73 | ||
More Information
Set Membership: |
|

PIP5K1A
Genomic Map