Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760168.3 | XP_002760214.1 | 3 - 4 | 458 | 73 | |
| XM_008984713.1 | XP_008982961.1 | 3 - 4 | 524 | 73 | ||
| XM_008984714.1 | XP_008982962.1 | 3 - 4 | 458 | 73 | ||
| XM_008984715.2 | XP_008982963.1 | 3 - 4 | 395 | 73 | ||
| XM_017966399.1 | XP_017821888.1 | 3 - 4 | 352 | 73 | ||
More Information
Set Membership: |
|

PEA15
Genomic Map