Assay Details
Gene Symbol: PI4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745938.3 | XP_002745984.1 | 4 - 5 | 937 | 90 | |
XM_008993519.2 | XP_008991767.1 | 4 - 5 | 937 | 90 | ||
XM_008993520.2 | XP_008991768.1 | 4 - 5 | 905 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
