Assay Details
Gene Symbol: ARPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003733239.2 | XP_003733287.1 | 7 - 8 | 731 | 60 | |
XM_008999470.1 | XP_008997718.1 | 7 - 8 | 640 | 60 | ||
XM_017973712.1 | XP_017829201.1 | 8 - 9 | 782 | 60 | ||
XM_017973713.1 | XP_017829202.1 | 6 - 7 | 588 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
