Assay Details
Gene Symbol: PPP1R9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008997386.2 | XP_008995634.1 | 9 - 10 | 2406 | 63 | |
XM_017972676.1 | XP_017828165.1 | 8 - 9 | 2301 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
