Assay Details
| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759386.3 | XP_002759432.1 | 2 - 3 | 359 | 91 | |
| XM_008983462.1 | XP_008981710.1 | 2 - 3 | 359 | 91 | ||
More Information
Set Membership: |
|

EIF5A2
Genomic Map