Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752934.4 | XP_002752980.2 | 8 - 9 | 634 | 94 | |
XM_017976332.1 | XP_017831821.1 | 3 - 4 | 483 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
