Assay Details
| Gene Symbol: GMDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746279.3 | XP_002746325.1 | 4 - 5 | 605 | 69 | |
| XM_017970947.1 | XP_017826436.1 | 4 - 5 | 605 | 69 | ||
More Information
Set Membership: |
|

GMDS
Genomic Map