Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752934.4 | XP_002752980.2 | 25 - 26 | 2733 | 88 | |
XM_009004526.2 | XP_009002774.1 | 24 - 25 | 2534 | 88 | ||
XM_017976332.1 | XP_017831821.1 | 20 - 21 | 2582 | 88 | ||
XM_017976333.1 | XP_017831822.1 | 18 - 19 | 2730 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
