Assay Details
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750803.3 | XP_002750849.1 | 1 - 2 | 890 | 66 | |
XM_017974712.1 | XP_017830201.1 | 1 - 2 | 363 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017974712.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
