Assay Details
Gene Symbol: NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003733355.3 | XP_003733403.2 | 2 - 3 | 1332 | 57 | |
XM_009000540.1 | XP_008998788.1 | 2 - 3 | 462 | 57 | ||
XM_009000541.2 | XP_008998789.1 | 3 - 4 | 528 | 57 | ||
XM_009000543.2 | XP_008998791.1 | 3 - 4 | 465 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
