Assay Details
Gene Symbol: CERS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749300.4 | XP_002749346.1 | 2 - 3 | 967 | 90 | |
XM_002749301.4 | XP_002749347.1 | 2 - 3 | 458 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
