Assay Details
Gene Symbol: CNTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008982045.2 | XP_008980293.1 | 8 - 9 | 1090 | 68 | |
XM_008982046.2 | XP_008980294.1 | 7 - 8 | 3300 | 68 | ||
XM_008982047.2 | XP_008980295.1 | 7 - 8 | 1562 | 68 | ||
XM_008982048.2 | XP_008980296.1 | 8 - 9 | 1087 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
