Assay Details
| Gene Symbol: HNF4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759010.3 | XP_002759056.3 | 9 - 10 | 2454 | 84 | |
| XM_008982803.2 | XP_008981051.1 | 10 - 11 | 1795 | 84 | ||
More Information
Set Membership: |
|

HNF4G
Genomic Map