Assay Details
| Gene Symbol: VDAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756193.4 | XP_002756239.1 | 5 - 6 | 1000 | 100 | |
| XM_017978976.1 | XP_017834465.1 | 4 - 5 | 419 | 100 | ||
More Information
Set Membership: |
|

VDAC2
Genomic Map