Assay Details
Gene Symbol: DEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003732583.3 | XP_003732631.1 | 3 - 4 | 689 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
