Assay Details
Gene Symbol: ANKMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002751540.4 | XP_002751586.1 | 7 - 8 | 1126 | 65 | |
XM_009002319.2 | XP_009000567.1 | 7 - 8 | 1126 | 65 | ||
XM_017975301.1 | XP_017830790.1 | 7 - 8 | 1126 | 65 | ||
XM_017975302.1 | XP_017830791.1 | 6 - 7 | 808 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
