Assay Details
| Gene Symbol: DTNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746206.3 | XP_002746252.2 | 3 - 4 | 576 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DTNBP1| Gene Symbol: DTNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746206.3 | XP_002746252.2 | 3 - 4 | 576 | 87 | |
Set Membership: |
|