Assay Details
| Gene Symbol: NR4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749430.4 | XP_002749476.2 | 4 - 5 | 1210 | 59 | |
| XM_003733180.1 | XP_003733228.1 | 4 - 5 | 1419 | 59 | ||
| XM_008998872.1 | XP_008997120.1 | 4 - 5 | 1419 | 59 | ||
More Information
Set Membership: |
|

NR4A2
Genomic Map