Assay Details
Gene Symbol: DAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745029.4 | XP_002745075.1 | 4 - 5 | 1058 | 105 | |
XM_008992132.2 | XP_008990380.1 | 5 - 6 | 1077 | 105 | ||
XM_008992133.2 | XP_008990381.1 | 4 - 5 | 863 | 105 | ||
XM_008992135.2 | XP_008990383.1 | 4 - 5 | 1058 | 105 | ||
XM_017969939.1 | XP_017825428.1 | 4 - 5 | 1058 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
