Assay Details
Gene Symbol: ARL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756561.3 | XP_002756607.1 | 2 - 3 | 310 | 109 | |
XM_009010205.1 | XP_009008453.1 | 2 - 3 | 214 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
