Assay Details
Gene Symbol: DELE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008986670.1 | XP_008984918.1 | 9 - 10 | 1243 | 70 | |
XM_008986676.1 | XP_008984924.1 | 9 - 10 | 1243 | 70 | ||
XM_008986688.1 | XP_008984936.1 | 9 - 10 | 1237 | 70 | ||
XM_008986693.1 | XP_008984941.1 | 8 - 9 | 1090 | 70 | ||
XM_017968447.1 | XP_017823936.1 | 9 - 10 | 1243 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
