Assay Details
Gene Symbol: NWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745924.3 | XP_002745970.1 | 5 - 6 | 916 | 76 | |
XM_008993495.2 | XP_008991743.1 | 5 - 6 | 942 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
