Assay Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753652.2 | XP_002753698.1 | 21 - 22 | 1966 | 66 | |
| XM_009005794.2 | XP_009004042.1 | 21 - 22 | 1949 | 66 | ||
| XM_009005795.2 | XP_009004043.1 | 21 - 22 | 1949 | 66 | ||
| XM_009005796.2 | XP_009004044.1 | 21 - 22 | 1948 | 66 | ||
| XM_009005797.2 | XP_009004045.1 | 21 - 22 | 1948 | 66 | ||
| XM_017977009.1 | XP_017832498.1 | 21 - 22 | 1949 | 66 | ||
| XM_017977010.1 | XP_017832499.1 | 21 - 22 | 1949 | 66 | ||
| XM_017977011.1 | XP_017832500.1 | 21 - 22 | 1950 | 66 | ||
| XM_017977012.1 | XP_017832501.1 | 21 - 22 | 1949 | 66 | ||
| XM_017977013.1 | XP_017832502.1 | 21 - 22 | 1952 | 66 | ||
More Information
Set Membership: |
|

CARMIL3
Genomic Map