Assay Details
Gene Symbol: LDB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756549.3 | XP_002756595.1 | 5 - 6 | 715 | 61 | |
XM_003734512.3 | XP_003734560.1 | 5 - 6 | 952 | 61 | ||
XM_009010192.2 | XP_009008440.1 | 5 - 6 | 715 | 61 | ||
XM_017979268.1 | XP_017834757.1 | 4 - 5 | 247 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
