Assay Details
Gene Symbol: NADK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745051.3 | XP_002745097.2 | 8 - 9 | 1062 | 71 | |
XM_008992163.2 | XP_008990411.1 | 9 - 10 | 1128 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
