Assay Details
| Gene Symbol: TRIM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744472.4 | XP_002744518.2 | 4 - 5 | 1189 | 58 | |
| XM_008989721.2 | XP_008987969.1 | 4 - 5 | 1154 | 58 | ||
| XM_008989729.2 | XP_008987977.1 | 4 - 5 | 1154 | 58 | ||
More Information
Set Membership: |
|

TRIM41
Genomic Map