Assay Details
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759807.3 | XP_002759853.1 | 3 - 4 | 842 | 88 | |
XM_017966187.1 | XP_017821676.1 | 2 - 3 | 730 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
