Assay Details
Gene Symbol: CNNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756510.3 | XP_002756556.1 | 5 - 6 | 2848 | 68 | |
XM_003734506.3 | XP_003734554.1 | 5 - 6 | 2849 | 68 | ||
XM_003734507.3 | XP_003734555.1 | 5 - 6 | 2849 | 68 | ||
XM_009010121.2 | XP_009008369.1 | 5 - 6 | 2850 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
