Assay Details
| Gene Symbol: ZNF384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975888.1 | XP_017831377.1 | 11 - 12 | 1690 | 91 | |
| XM_017975889.1 | XP_017831378.1 | 11 - 12 | 1749 | 91 | ||
| XM_017975890.1 | XP_017831379.1 | 11 - 12 | 1758 | 91 | ||
| XM_017975891.1 | XP_017831380.1 | 12 - 13 | 1841 | 91 | ||
| XM_017975892.1 | XP_017831381.1 | 12 - 13 | 1732 | 91 | ||
| XM_017975893.1 | XP_017831382.1 | 10 - 11 | 1642 | 91 | ||
| XM_017975894.1 | XP_017831383.1 | 10 - 11 | 1701 | 91 | ||
| XM_017975895.1 | XP_017831384.1 | 11 - 12 | 1793 | 91 | ||
| XM_017975897.1 | XP_017831386.1 | 10 - 11 | 1646 | 91 | ||
| XM_017975898.1 | XP_017831387.1 | 9 - 10 | 1549 | 91 | ||
| XM_017975899.1 | XP_017831388.1 | 9 - 10 | 2014 | 91 | ||
| XM_017975900.1 | XP_017831389.1 | 9 - 10 | 1414 | 91 | ||
| XM_017975901.1 | XP_017831390.1 | 7 - 8 | 1873 | 91 | ||
More Information
Set Membership: |
|

ZNF384
Genomic Map