Assay Details
| Gene Symbol: ACVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749420.3 | XP_002749466.1 | 10 - 11 | 1935 | 68 | |
| XM_002749421.3 | XP_002749467.1 | 10 - 11 | 1643 | 68 | ||
| XM_008998867.2 | XP_008997115.1 | 9 - 10 | 1494 | 68 | ||
| XM_017973469.1 | XP_017828958.1 | 8 - 9 | 1490 | 68 | ||
More Information
Set Membership: |
|

ACVR1
Genomic Map