Assay Details
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002748480.3 | XP_002748526.1 | 4 - 5 | 1473 | 67 | |
XM_002748481.3 | XP_002748527.1 | 3 - 4 | 1383 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
