Assay Details
| Gene Symbol: SUPT3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994522.2 | XP_008992770.1 | 6 - 7 | 767 | 109 | |
| XM_008994523.2 | XP_008992771.1 | 5 - 6 | 482 | 109 | ||
More Information
Set Membership: |
|

SUPT3H
Genomic Map