Assay Details
| Gene Symbol: NR1H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735633.3 | XP_003735681.3 | 5 - 6 | 742 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR1H2| Gene Symbol: NR1H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735633.3 | XP_003735681.3 | 5 - 6 | 742 | 74 | |
Set Membership: |
|