Assay Details
| Gene Symbol: AKAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748063.2 | XP_002748109.1 | 12 - 13 | 2001 | 73 | |
| XM_008996761.2 | XP_008995009.1 | 11 - 12 | 1832 | 73 | ||
| XM_017972409.1 | XP_017827898.1 | 12 - 13 | 2387 | 73 | ||
More Information
Set Membership: |
|

AKAP10
Genomic Map