Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760491.4 | XP_002760537.1 | 2 - 3 | 1044 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP10| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760491.4 | XP_002760537.1 | 2 - 3 | 1044 | 87 | |
Set Membership: |
|